Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

86848

Sigma-Aldrich

Tetrabutylammonium ethoxide solution

~40% in ethanol (NT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(OCH2CH3)
CAS Number:
Molecular Weight:
287.52
Beilstein:
6018401
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

~40% in ethanol (NT)

storage temp.

2-8°C

SMILES string

CC[O-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.C2H5O/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-2-3/h5-16H2,1-4H3;2H2,1H3/q+1;-1

InChI key

SNUBRLWUQHFANK-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

73.4 °F

Flash Point(C)

23 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service