Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

86334

Sigma-Aldrich

TentaGel HL-NH2

extent of labeling: ~0.40 mmol/g loading

Synonym(s):

O-(2-Aminoethyl)polyethylene glycol polymer-bound

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 G
$311.00

$311.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
2.5 G
$311.00

About This Item

UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$311.00


Please contact Customer Service for Availability

Request a Bulk Order

product line

TentaGel

form

solid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

~0.40 mmol/g loading

particle size

~110 μm

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

Legal Information

TentaGel is a trademark of Rapp Polymere GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service