Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

862185

Sigma-Aldrich

3-(4-Hydroxyphenyl)propionic acid N-hydroxysuccinimide ester

90%, technical grade

Synonym(s):

N-Succinimidyl 3-(4-hydroxyphenyl)propionate, N-[(4-Hydroxyhydrocinnamoyl)oxy]succinimide, Bolton-Hunter reagent precursor, Rudinger reagent, SHPP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13NO5
CAS Number:
Molecular Weight:
263.25
Beilstein:
1545011
EC Number:
MDL number:
UNSPSC Code:
12352000

grade

technical grade

Assay

90%

impurities

10% isopropyl ether

mp

132-133 °C (lit.)

SMILES string

Oc1ccc(CCC(=O)ON2C(=O)CCC2=O)cc1

InChI

1S/C13H13NO5/c15-10-4-1-9(2-5-10)3-8-13(18)19-14-11(16)6-7-12(14)17/h1-2,4-5,15H,3,6-8H2

InChI key

KYRUKRFVOACELK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service