Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

86141

Sigma-Aldrich

Dicyclohexyl sulfosuccinate sodium salt

≥98.0% (TLC)

Synonym(s):

Dicyclohexyl sodium sulfosuccinate, Sodium dicyclohexyl sulfosuccinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H25NaO7S
CAS Number:
Molecular Weight:
384.42
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

description

anionic

Assay

≥98.0% (TLC)

form

fibers

mol wt

384.42 g/mol

impurities

≤2% water

application(s)

detection

functional group

ester
sulfonic acid

SMILES string

[Na+].[O-]S(=O)(=O)C(CC(=O)OC1CCCCC1)C(=O)OC2CCCCC2

InChI

1S/C16H26O7S.Na/c17-15(22-12-7-3-1-4-8-12)11-14(24(19,20)21)16(18)23-13-9-5-2-6-10-13;/h12-14H,1-11H2,(H,19,20,21);/q;+1/p-1

InChI key

UELAIMNOXLAYRW-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service