Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

860832

Sigma-Aldrich

2-Thiocytidine dihydrate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N3O4S · 2H2O
CAS Number:
Molecular Weight:
295.31

Assay

98%

SMILES string

NC1=NC(=S)N(C=C1)[C@H]2O[C@H](CO)[C@@H](O)[C@H]2O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Wang et al.
Nucleic acids research, 25(21), 4355-4361 (1997-10-23)
The hammerhead domain is one of the smallest known ribozymes. Like other ribozymes it catalyzes site-specific cleavage of a phosphodiester bond. The hammerhead ribozyme has been the subject of a vast number of biochemical and structural studies aimed at determining
Akihiro Ohkubo et al.
Organic & biomolecular chemistry, 10(10), 2008-2010 (2012-01-28)
In this paper, we report DNA and 2'-OMe-RNA probes containing 5-methyl-2-thiocytidine (m(5)s(2)C) residues that can bind selectively and strongly to the corresponding RNA targets containing inosine residues by the significant stacking effect and steric hindrance of the 2-thiocarbonyl group.
D J Earnshaw et al.
Biochemistry, 39(21), 6410-6421 (2000-06-01)
The hairpin ribozyme achieves catalytic cleavage through interaction of essential nucleotides located in two distinct helical domains that include internal loops. Initial docking of the two domains is ion dependent and appears to be followed by a structural rearrangement that
R L Joshi et al.
FEBS letters, 208(2), 189-193 (1986-11-24)
A new approach for the fluorescence labeling of an aminoacyl-tRNA at the 3'-end is applied to study its interaction with bacterial elongation factor Tu (EF-Tu) and GTP at equilibrium. The penultimate cytidine residue in yeast tRNATyr-C-C-A was replaced by 2-thiocytidine
T Döring et al.
The EMBO journal, 13(11), 2677-2685 (1994-06-01)
A photo-reactive diazirine derivative was attached to the 2-thiocytidine residue at position 32 of tRNA(Arg)I from Escherichia coli. This modified tRNA was bound under suitable conditions to the A, P or E site of E.coli ribosomes. After photo-activation of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service