Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

860522

Sigma-Aldrich

Methyl β-D-arabinopyranoside

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O5
CAS Number:
Molecular Weight:
164.16
EC Number:
MDL number:
UNSPSC Code:
12352103

Quality Level

Assay

99%

SMILES string

O[C@@H]1CO[C@@H](O)[C@@H](O)[C@@H]1O

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Goeminne et al.
European journal of medicinal chemistry, 43(2), 315-326 (2007-06-22)
Nucleoside hydrolase (NH) is a key enzyme in the purine salvage pathway. The purine specificity of the IAG-NH from Trypanosoma vivax is at least in part due to cation-pi-stacking interactions. Guanidinium ions can be involved in cation-pi-stacking interactions, therefore a
Jiyoung A Hong et al.
Journal of medicinal chemistry, 52(17), 5485-5495 (2009-08-15)
Mycobacterium tuberculosis adenosine-5'-phosphosulfate (APS) reductase is an iron-sulfur protein and a validated target to develop new antitubercular agents, particularly for the treatment of latent infection. To facilitate the development of potent and specific inhibitors of APS reductase, we have probed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service