859362
L-Histidinol dihydrochloride
99%
Synonym(s):
β-Aminoimidazole-4-propanol dihydrochloride, (S)-2-Amino-3-(4-imidazolyl)propanol dihydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H11N3O · 2HCl
CAS Number:
Molecular Weight:
214.09
Beilstein:
3914853
EC Number:
MDL number:
UNSPSC Code:
12352103
Assay
99%
optical activity
[α]19/D −2.2°, c = 2 in H2O
mp
198-201 °C (dec.) (lit.)
SMILES string
Cl[H].Cl[H].N[C@H](CO)Cc1c[nH]cn1
InChI
1S/C6H11N3O.2ClH/c7-5(3-10)1-6-2-8-4-9-6;;/h2,4-5,10H,1,3,7H2,(H,8,9);2*1H/t5-;;/m0../s1
InChI key
FRCAFNBBXRWXQA-XRIGFGBMSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Precursor of histamine; reversible inhibitor of protein synthesis.
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service