Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

859036

Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl β-D-glucopyranoside

98%

Synonym(s):

X-Glc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15BrClNO6
CAS Number:
Molecular Weight:
408.63
Beilstein:
1552603
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

SMILES string

OC[C@H]1O[C@@H](Oc2c[nH]c3ccc(Br)c(Cl)c23)[C@H](O)[C@@H](O)[C@@H]1O

Application

For the histochemical determination of β-glucosidase.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biol. Plant., 15, 88-88 (1973)
W D Watkins et al.
Applied and environmental microbiology, 54(7), 1874-1875 (1988-07-01)
A new chromogenic compound, 5-bromo-4-chloro-3-indoxyl-beta-D-glucuronide, was found to be useful for the rapid, specific, differential identification of Escherichia coli in the sanitary analysis of shellfish and wastewater. Of 1,025 presumptively positive colonies (blue) and 583 presumptively negative colonies (nonblue), only
L A Shelef et al.
Letters in applied microbiology, 25(3), 202-206 (1997-11-14)
A new instrument, capable of detecting metabolic changes due to microbiological activity, is described. Optical changes in growth media are monitored in a semi-fluid zone that separates the liquid medium containing the sample. Data demonstrate that common media can be
R Gossrau et al.
Histochemistry, 86(4), 397-404 (1987-01-01)
5-Br-4-Cl-3-Indoxyl-alpha-D-gluco(pyrano)side was found to be the most suitable synthetic substrate for the demonstration of alpha-D-glucosidases in situ. Using an azoindoxyl procedure with hexazotized pararosaniline or new fuchsine at pH 5 in freeze-dried celloidine-mounted cryostat sections acid alpha-D-glucosidase (EC 3.2.1.20) was
H Arakawa et al.
Analytical biochemistry, 199(2), 238-242 (1991-12-01)
Chemiluminescent assays of various enzymes have been developed using indoxyl derivatives as substrates. The principle of the method is as follows: an enzyme causes hydrolysis of an indoxyl derivative to an intermediate indoxyl that is readily oxidized to indigo dye

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service