Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

857823

Sigma-Aldrich

4-Sulfophenyl isothiocyanate sodium salt monohydrate

technical grade

Synonym(s):

4-Isothiocyanatobenzenesulfonic acid sodium salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaO3SC6H4NCS·H2O
CAS Number:
Molecular Weight:
255.25
Beilstein:
3777198
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

form

powder

functional group

isothiocyanate
sulfonic acid

storage temp.

2-8°C

SMILES string

[Na+].[H]O[H].[O-]S(=O)(=O)c1ccc(cc1)N=C=S

InChI

1S/C7H5NO3S2.Na.H2O/c9-13(10,11)7-3-1-6(2-4-7)8-5-12;;/h1-4H,(H,9,10,11);;1H2/q;+1;/p-1

InChI key

TYLQYONISARSPL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service