Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

855634

Sigma-Aldrich

N-Acetyl-4-fluoro-DL-phenylalanine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2CH(NHCOCH3)CO2H
CAS Number:
Molecular Weight:
225.22
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

form

crystals

reaction suitability

reaction type: solution phase peptide synthesis

mp

150-153 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(=O)NC(Cc1ccc(F)cc1)C(O)=O

InChI

1S/C11H12FNO3/c1-7(14)13-10(11(15)16)6-8-2-4-9(12)5-3-8/h2-5,10H,6H2,1H3,(H,13,14)(H,15,16)

InChI key

NRLBRFQYMSTLJK-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service