Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

855553

Sigma-Aldrich

β-Carotene

95%

Synonym(s):

trans-β-Carotene, Provitamin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H56
CAS Number:
Molecular Weight:
536.87
Beilstein:
1917416
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

95%

mp

178-179 °C (lit.)

λmax

450 nm
478 nm (2nd)

SMILES string

CC1=C(\C=C\C(C)=C\C=C\C(C)=C\C=C\C=C(C)\C=C\C=C(C)\C=C\C2=C(C)CCCC2(C)C)C(C)(C)CCC1

Application

Provitamin A1. Antioxidant in biological systems.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Zamora et al.
The Journal of nutrition, 121(1), 50-56 (1991-01-01)
Five groups of five weanling rats were each fed a Torula yeast-based diet either unsupplemented or supplemented with 30 mg beta-carotene/kg, 30 IU vitamin E/kg, 1 mg selenium/kg or 30 mg coenzyme Q10/kg. Elevated levels of plasma aspartate aminotransferase and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service