Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

855472

Sigma-Aldrich

S-Methyl-L-cysteine

97%

Synonym(s):

(R)-2-Amino-3-(methylmercapto)propionic acid, SMLC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
135.18
Beilstein:
1721675
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

97%

optical activity

[α]20/D −30°, c = 1 in H2O

SMILES string

CSC[C@H](N)C(O)=O

InChI

1S/C4H9NO2S/c1-8-2-3(5)4(6)7/h3H,2,5H2,1H3,(H,6,7)/t3-/m0/s1

InChI key

IDIDJDIHTAOVLG-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service