Skip to Content
MilliporeSigma
All Photos(1)

Documents

852910

Sigma-Aldrich

4-Aminobenzoic acid sodium salt

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CO2Na
CAS Number:
Molecular Weight:
161.13
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

99%

mp

>300 °C (lit.)

SMILES string

[Na+].Nc1ccc(cc1)C([O-])=O

InChI

1S/C7H7NO2.Na/c8-6-3-1-5(2-4-6)7(9)10;/h1-4H,8H2,(H,9,10);/q;+1/p-1

InChI key

XETSAYZRDCRPJY-UHFFFAOYSA-M

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Todd A Smith et al.
Genome biology and evolution, 7(3), 831-838 (2015-01-27)
Amblyomma americanum (Lone star tick) is an important disease vector in the United States. It transmits several human pathogens, including the agents of human monocytic ehrlichiosis, tularemia, and southern tick-associated rash illness. Blood-feeding insects (Class Insecta) depend on bacterial endosymbionts
Yasmin Mossavar-Rahmani et al.
American journal of epidemiology, 181(12), 996-1007 (2015-05-23)
We investigated measurement error in the self-reported diets of US Hispanics/Latinos, who are prone to obesity and related comorbidities, by background (Central American, Cuban, Dominican, Mexican, Puerto Rican, and South American) in 2010-2012. In 477 participants aged 18-74 years, doubly
Ashley R Heurung et al.
Dermatitis : contact, atopic, occupational, drug, 25(6), 289-326 (2014-11-11)
Sunscreen is a key component in the preventive measures recommended by dermatologists and public health campaigns aimed at reducing sunburn, early skin aging, and skin cancer. To maximize compliance, adverse reactions to sunscreens should be minimized. Although inactive ingredients cause
Brian L Henry et al.
Thrombosis research, 134(5), 1123-1129 (2014-09-23)
Sulfated low molecular weight lignins (LMWLs) have been found to bind in the heparin binding sites of coagulation proteinases. LMWLs represent a library of diverse non-carbohydrate, aromatic molecules which are structures different from heparin, but still potently inhibit thrombin and
Carlos E Rodríguez-Rodríguez et al.
Bioresource technology, 168, 180-189 (2014-03-04)
The use of Trametes versicolor has been partially successful in the removal of some pharmaceuticals from sewage sludge in laboratory-scale biopile systems. The application of two strategies for the re-inoculation of biomass was assessed during the fungal bioaugmentation of non-sterile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service