Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

852627

Sigma-Aldrich

2,6-Dithiopurine

Synonym(s):

2,6-Dimercaptopurine, 2,6-Purinedithiol, Dithioxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N4S2
CAS Number:
Molecular Weight:
184.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

mp

>350 °C (lit.)

SMILES string

Sc1nc(S)c2[nH]cnc2n1

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M C MacLeod et al.
Cancer research, 51(18), 4859-4864 (1991-09-25)
The chemotherapeutic agent 6-mercaptopurine was previously shown to inhibit the binding of 7r,8t-dihydroxy-9,10t-oxy-7,8,9,10-tetrahydro-benzo(a) pyrene (BPDE-I) to DNA in Chinese hamster ovary cells. Two compounds related to 6-mercaptopurine, 2,6-dithiopurine (DTP) and thiopurinol (TP), have been tested for inhibition of the binding
K Datta et al.
Toxicology, 125(1), 1-11 (1998-05-19)
2,6-Dithiopurine (DTP) has been proposed as a possible chemopreventive agent because of its ability to react with electrophiles. Acrolein, an electrophilic metabolite of cyclophosphamide (CP) involved in the toxicities of this anticancer drug, can be scavenged by DTP. The present
W G Qing et al.
Drug metabolism and disposition: the biological fate of chemicals, 23(8), 854-860 (1995-08-01)
2,6-Dithiopurine (DTP) has been proposed as a possible chemopreventive agent because of its facile reaction with the electrophilic ultimate carcinogen, benzo[a]pyrene diol epoxide, and other reactive electrophiles. Previous studies in mouse skin indicated almost complete inhibition of benzo[a]pyrene diol epoxide-induced
Stephen Boulware et al.
Toxicology and applied pharmacology, 263(2), 203-209 (2012-06-27)
Sulfur mustard [bis(2-chloroethyl)sulfide, SM] is a well-known DNA-damaging agent that has been used in chemical warfare since World War I, and is a weapon that could potentially be used in a terrorist attack on a civilian population. Dermal exposure to
W G Qing et al.
Chemical research in toxicology, 9(8), 1298-1304 (1996-12-01)
Purinethiols are a class of potential cancer chemopreventive agents that exhibit nucleophilic scavenging activity against the carcinogenic electrophile benzo[a]pyrene diol epoxide (BPDE). Of the purinethiols tested previously, 2,6-dithiopurine (DTP), exhibited the highest scavenging activity for BPDE when tested either in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service