Skip to Content
MilliporeSigma
All Photos(1)

Documents

851671

Sigma-Aldrich

Helicin

99%

Synonym(s):

Salicylaldehyde β-D-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16O7
CAS Number:
Molecular Weight:
284.26
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

optical activity

[α]25/D −57.1°, c = 1.5 in H2O

mp

178 °C (lit.)

SMILES string

[H]C(=O)c1ccccc1O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O

InChI

1S/C13H16O7/c14-5-7-3-1-2-4-8(7)19-13-12(18)11(17)10(16)9(6-15)20-13/h1-5,9-13,15-18H,6H2/t9-,10-,11+,12-,13-/m1/s1

InChI key

BGOFCVIGEYGEOF-UJPOAAIJSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Line Christiansen et al.
Applied and environmental microbiology, 86(21) (2020-08-23)
Pseudomonas fluorescens In5 synthesizes the antifungal cyclic lipopeptides (CLPs) nunamycin and nunapeptin, which are similar in structure and genetic organization to the pseudomonas-derived phytotoxins syringomycin and syringopeptin. Regulation of syringomycin and syringopeptin is dependent on the two-component global regulatory system

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service