Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

850624

Sigma-Aldrich

Hydantoin-5-acetic acid

98%

Synonym(s):

2,4-Dioxoimidazolidine-5-acetic acid, 5-Hydantoinacetic acid, DL-5-(Carboxymethyl)hydantoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O4
CAS Number:
Molecular Weight:
158.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

214-215 °C (dec.) (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)CC1NC(=O)NC1=O

InChI

1S/C5H6N2O4/c8-3(9)1-2-4(10)7-5(11)6-2/h2H,1H2,(H,8,9)(H2,6,7,10,11)

InChI key

DQQLZADYSWBCOX-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Nonfolate compounds as antiparasitic agents inhibiting pteridine reductase
  • 2,4-Azolidinedione-acetic acid derivatives as anticancer agents
  • Anti-inflammatory and analgesic drugs
  • Amides via polystyrene-supported N-hydroxybenxotriazole resin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Chylińska et al.
Molecules (Basel, Switzerland), 25(16) (2020-08-23)
Current demand for new protective materials ensuring sterility is systematically growing. The purpose of this work was the synthesis of the biocidal N-halamine hydantoin-containing chitosan (CS-CMH-Cl) and characterization of its properties. The functionalization of the chitosan by 5-hydantoinacetic acid substitution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service