Skip to Content
MilliporeSigma

850071

Sigma-Aldrich

Glycyl-L-leucine

99%

Synonym(s):

Gly-Leu

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CONHCH(COOH)CH2CH(CH3)2
CAS Number:
Molecular Weight:
188.22
Beilstein:
1726294
EC Number:
MDL number:
UNSPSC Code:
12352103

Pricing and availability is not currently available.

Assay

99%

optical activity

[α]22/D −34.5°, c = 2 in H2O

mp

233-235 °C (lit.)

SMILES string

CC(C)C[C@H](NC(=O)CN)C(O)=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service