Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

84656

Sigma-Aldrich

1,3-Di-Boc-2-(carboxymethyl)guanidine

≥97.0%

Synonym(s):

N-(N,N′-Di-Boc-guanyl)glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H23N3O6
CAS Number:
Molecular Weight:
317.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97.0%

form

powder

color

white

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)NC(NC(=O)OC(C)(C)C)=NCC(O)=O

InChI

1S/C13H23N3O6/c1-12(2,3)21-10(19)15-9(14-7-8(17)18)16-11(20)22-13(4,5)6/h7H2,1-6H3,(H,17,18)(H2,14,15,16,19,20)

InChI key

ZJYJFQDYIMDGDG-UHFFFAOYSA-N

Other Notes

Guanidinylation reagent

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K C Nicolaou et al.
Bioorganic & medicinal chemistry, 6(8), 1185-1208 (1998-10-24)
Recent studies demonstrated that peptide and antibody antagonists of integrin alpha v beta 3 block angiogenesis and tumor growth. In this article, the design, synthesis and biological evaluation of a series of nitroaryl ether-based, nonpeptide mimetics are described. The design

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service