Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

84590

Sigma-Aldrich

(+)-Sativene

≥98.0% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein:
6566323
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (sum of enantiomers, GC)

optical activity

[α]20/D +176±3°, c = 10% in ethanol

refractive index

n20/D 1.496

bp

256 °C (lit.)

density

0.921 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

[H][C@]12CC[C@]3([H])[C@@]1([H])[C@@H](CC[C@]3(C)C2=C)C(C)C

InChI

1S/C15H24/c1-9(2)11-7-8-15(4)10(3)12-5-6-13(15)14(11)12/h9,11-14H,3,5-8H2,1-2,4H3/t11-,12+,13+,14-,15+/m0/s1

InChI key

VOBBUADSYROGAT-VYDRJRHOSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service