Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

83886

Sigma-Aldrich

Rink amide (aminomethyl)polystyrene with Norleucine internal standard

extent of labeling: ~1.1 mmol/g loading

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

~1.1 mmol/g loading

matrix

crosslinked with 1% DVB

particle size

200-400 mesh

application(s)

peptide synthesis

functional group

amine

Application

  • Solid support for Fmoc-based solid phase peptide synthesis (SPPS) of peptide amides.
  • This solid support contains a modified Rink amide linker attached through norleucine to aminomethylpolystyrene resin.
  • Cleavage of peptides is achieved by adding 95% TFA.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service