Skip to Content
MilliporeSigma
All Photos(1)

Documents

83720

Sigma-Aldrich

Rhodium on activated alumina

5% rhodium (Rh) basis

Synonym(s):

Rh

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: rhodium
reagent type: catalyst

concentration

5% Rh

Application

Catalyst for hydrogenation of unsaturated compounds including aldehydes, ketones, andaromatic carbocyclic and heterocyclic structures; high stereospecificity and low hydrogenolytic activity.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service