Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8300NG6

Sigma-Aldrich

Conjugated Polymer Nanoparticle (CPN)

fluorescence λem 830 nm (IR-I), carboxy-NHS-amine functionalized, 1 ea of 30 μg antibody kit

Synonym(s):

CPN 30-minute conjugation kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$472.00

$472.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$472.00

About This Item

UNSPSC Code:
12162002
NACRES:
NA.51

$472.00


Please contact Customer Service for Availability

Request a Bulk Order

packaging

1 ea of 30 μg antibody kit

fluorescence

λem 830 nm (IR-I)

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

CPNs are extremely bright and stable fluorophore nanoparticles which have been shown to give exceptional performance in a wide range of fluorescent analysis applications, such as lateral flow tests, fluorescent ELISA, cell tracking, immunofluorescence microscopy, flow cytometry and western blot. CPNs are resistant to photobleaching and a wide range of temperatures (-20°C to 120°C) and pH (2 to 12). They have been shown to be 1000x brighter than other fluorophores bringing exceptional levels of detection to assays and diagnostics.

Application

Amine Antibody Conjugation kit allows covalent linkage reactions to be readily carried out without access to specialist chemistry capabilities. The kit is designed to be specific for CPN – antibody / protein linkage has a rapid reaction time of 30mins and comes in three sizes for the conjugation of 10mcg, 30mcg and 100mcg of antibodies. CPNs are available in a range of emission wavelengths covering the visible and IR spectrum.

Legal Information

CPN is a trademark of Stream Bio Ltd

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service