Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

81964

Sigma-Aldrich

1-Boc-4-piperidylacetic acid

≥98.0% (TLC)

Synonym(s):

1-Boc-4-piperidineacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H21NO4
CAS Number:
Molecular Weight:
243.30
Beilstein:
7028968
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98.0% (TLC)

form

crystals

reaction suitability

reaction type: Boc solid-phase peptide synthesis

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N1CCC(CC1)CC(O)=O

InChI

1S/C12H21NO4/c1-12(2,3)17-11(16)13-6-4-9(5-7-13)8-10(14)15/h9H,4-8H2,1-3H3,(H,14,15)

InChI key

ZXFLMSIMHISJFV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service