Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

81560

Sigma-Aldrich

Polystyrene A-RAM

extent of labeling: 0.4-0.7 mmol/g loading

Synonym(s):

Rink amide(2-aminoethyl)polystyrene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

beads

extent of labeling

0.4-0.7 mmol/g loading

matrix

crosslinked with 1% DVB

particle size

500-560 μm

application(s)

peptide synthesis

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan Zhang et al.
Clinica chimica acta; international journal of clinical chemistry, 489, 203-211 (2017-12-11)
Accumulated data indicate that placental hypoxia is implicated in the pathogenesis of preeclampsia (PE). Tight junction (TJ) is important structure that sustains normal placental barrier function, its dysregulation under hypoxia has been observed. This study was designed to explore hypoxia-induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service