Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

80982

Sigma-Aldrich

Platinum on activated charcoal

greener alternative

5% Pt basis

Synonym(s):

Pt/C

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

solid

reaction suitability

core: platinum
reagent type: catalyst

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

5% Pt

matrix

activated charcoal support

greener alternative category

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Platinum on activated carbon (Pt/C) can be used as a catalyst for a variety of applications such as:
  • hydrogenation of D-fructose and D-glucose
  • hydrosilylation reaction for the production of molecules with silicon-carbon bonds
  • direct alcohol fuel cells (DAFCs)

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrogenation of d-fructose over activated charcoal supported platinum catalyst
Ahmed MJ and Kadhum AAH
Journal of the Taiwan Institute of Chemical Engineers, 42(1), 114-119 (2011)
Kinetics studies of d-glucose hydrogenation over activated charcoal supported platinum catalyst
Ahmed MJ
Chemosphere, 48(2), 343-347 (2012)
Ethanol electrooxidation on Pt/C and Pd/C catalysts promoted with oxide
Xu C, et al.
Journal of Power Sources, 164(2), 527-531 (2007)
Ethanol oxidation reactions using SnO2@ Pt/C as an electrocatalyst
Silva JCM, et al.
Applied Catalysis. B, Environmental, 99(1-2), 265-271 (2010)
Hydrosilylation of alkynes catalyzed by platinum on carbon
Chauhan M, et al.
Journal of Organometallic Chemistry, 645(1-2), 1-13 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service