Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

809241

Sigma-Aldrich

N-Ethyl-N-isopropylpropan-2-aminium 2,2-dimethyl-4-oxo-5-(perfluoropyridin-4-yl)-4H-1,3-dioxin-6-olate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26F4N2O4
CAS Number:
Molecular Weight:
422.41
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

powder

mp

100-105 °C

storage temp.

2-8°C

Application

N-Ethyl-N-isopropylpropan-2-aminium 2,2-dimethyl-4-oxo-5-(perfluoropyridin-4-yl)-4H-1,3-dioxin-6-olate is a compound undergoes cycloreversion to form the fluoro(hetero)aryl ketene, which undergoes efficient coupling with nucleophiles and allows rapid incorporation of highly fluorinated α-fluoro(hetero)aryl acetic acid derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Organofluorine chemistry is an essential part of drug discovery programs as well as agrochemical programs and even plays a major role in materials chemistry. Despite the undeniable importance of fluorinated organic molecules, our ability to synthesize these substrates is lacking - though arguably it is better than that of Nature. Consequently, methods that allow facile access to fluorinated molecules are important especially when they provide unique access to fluorinated chemical space.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service