Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

808202

Sigma-Aldrich

Zinc oxide nanoparticle ink

2.5 wt. %, viscosity 10 cP, work function -3.9eV

Synonym(s):

Avantama N-11-Jet, Nanograde N-11-Jet, ZnO ink, ZnO nanoparticle ink, zinc oxide dispersion, zinc oxide suspension

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

form

dispersion

concentration

2.5 wt. % (crystalline ZnO in isopropanol and propylene glycol)

work function

-3.7--4.1 eV

particle size

8-16 nm

viscosity

8-14 cP

Looking for similar products? Visit Product Comparison Guide

Application

ZnO nanoparticle ink is for inkjet printing for the use as electron transport layer in printed electronics. ZnO nanoparticle ink is universally applicable in normal and inverted architecture.

Preparation Note

  • Storage: In dark at room temperature.
  • Prior to application: Shake, ultrasonicate with sonic horn and(optionally) filter through 0.45μm PTFE filter.
  • Post-treatment: Annealing of deposited ZnO films at >120°C.

Legal Information

Product of Avantama Ltd.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Progress in solution-processed functional materials leads to thin-film optoelectronic devices for industrial and consumer electronics.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service