Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

808172

Sigma-Aldrich

Aluminum-doped zinc oxide nanoparticle ink

2.5 wt. %, viscosity 11 cP, work function -4.3eV

Synonym(s):

AZO ink, Al-doped ZnO dispersion, Al-dopend ZnO suspension, Al:ZnO ink, Avantama N-20X-Jet, Nanograde N-20X-Jet

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

form

dispersion

concentration

2.5 wt. % (crystalline Al doped ZnO (3.15 mol% Al) in 2-propanol and propylene glycol)

work function

-4.1--4.5 eV

particle size

8-16 nm

viscosity

8-14 cP

Looking for similar products? Visit Product Comparison Guide

Application

Al-doped ZnO nanoparticle ink is for slot dye or inkjet printing for the use as electron transport layer in printed electronics. Al-doped ZnO nanoparticle ink is universally applicable in normal and inverted architecture with performance comparable to evaporated calcium.

Preparation Note

  • Storage: In dark at room temperature.
  • Prior to application: Shake, ultrasonicate with sonic horn and (optionally) filter through 0.45 μm PTFE filter.
  • Post-treatment: Annealing of deposited Al doped ZnO films at >100°C.

Legal Information

Product of Avantama Ltd.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

73.4 °F

Flash Point(C)

23 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Progress in solution-processed functional materials leads to thin-film optoelectronic devices for industrial and consumer electronics.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service