Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

806471

Sigma-Aldrich

Poly(N-isopropylacrylamide)

average Mn 30,000

Synonym(s):

NIPAM polymer, PNIPAM, Polyacrylamide, polyNIPAM

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(C6H11NO)nCH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

mol wt

average Mn 30,000

storage temp.

2-8°C

SMILES string

CC(C)NC(=O)C=C

InChI

1S/C6H11NO/c1-3-5-7-6(8)4-2/h4H,2-3,5H2,1H3,(H,7,8)

InChI key

WDFKEEALECCKTJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

PolyNIPAM is a stimuli-responsive polymer. The low PDI means that the distribution of chain lengths is narrow which will typically lead to better reproducibility. This polymer for development of thermosensitive coated micro/nano materials, including thermoresponsive polymeric drug delivery systems.
Thermosensitive polymer, can be used to form a hydrogel. Aqueous polymer solution undergoes a phase transition from a soluble to an insoluble state when the temperature is raised to ca. 32 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Professor Mitsuhiro Ebara provides insights on several types of smart nanofiber mesh systems that have been explored for different drug delivery purposes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service