Skip to Content
MilliporeSigma
All Photos(1)

Documents

806005

Sigma-Aldrich

APN-Amine HCl

95%

Synonym(s):

3-(4-(4-(Aminomethyl)-1H-1,2,3-triazol-1-yl)phenyl)propiolonitrile hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10N5Cl
Molecular Weight:
259.69
UNSPSC Code:
12352200
PubChem Substance ID:

Quality Level

Assay

95%

form

powder

reaction suitability

reagent type: cross-linking reagent

storage temp.

2-8°C

SMILES string

N#CC#CC1=CC=C(N2C=C(CN)N=N2)C=C1.[H]Cl

InChI

1S/C12H9N5.ClH/c13-7-1-2-10-3-5-12(6-4-10)17-9-11(8-14)15-16-17;/h3-6,9H,8,14H2;1H

InChI key

JZGAUVVBWJHLGN-UHFFFAOYSA-N

Application

APN-Amine is a bifunctional crosslinker for thiol-to-carboxyl coupling. The coupling can be performed with high selectivity in biological medium using mild reaction conditions. The amino group readily reacts with activated carboxylic acids (e.g. NHS-esters) in basic conditions to produce stable amide bonds. The APN group can be used for coupling with free thiols (e.g. labeling of free cysteines in proteins).

Other Notes

Standard protein labeling procedure (cysteine labeling):
  • Dissolve the protein in the appropriate buffer* with pH 6.5-9.0 (e.g. PBS) at 1-10 mg/mL concentration.
  • Apply the appropriate amount of the stock solution of the reagent (1-5 molar eq. per free cysteine residue).
  • Incubate at room temperature for 2 hours.
  • If necessary, purify the protein conjugate using size exclusion chromatography or ultrafiltration.
*Note: avoid thiol-containing buffers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergii Kolodych et al.
Bioconjugate chemistry, 26(2), 197-200 (2015-01-24)
Amine-to-thiol coupling is the most common route for the preparation of antibody-drug conjugates (ADC). It is usually achieved by using heterobifunctional reagents possessing an activated ester at one end and a maleimide group at the other. However, maleimide-based conjugates were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service