Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80540

Sigma-Aldrich

Pinacryptol yellow

for photographic purposes

Synonym(s):

6-Ethoxy-1-methyl-2-(3-nitro-β-styryl)quinolinium methyl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22N2O7S
CAS Number:
Molecular Weight:
446.47
Beilstein:
3881592
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

mp

260-262 °C (lit.)

SMILES string

COS([O-])(=O)=O.CCOc1ccc2[n+](C)c(\C=C\c3cccc(c3)[N+]([O-])=O)ccc2c1

InChI

1S/C20H19N2O3.CH4O4S/c1-3-25-19-11-12-20-16(14-19)8-10-17(21(20)2)9-7-15-5-4-6-18(13-15)22(23)24;1-5-6(2,3)4/h4-14H,3H2,1-2H3;1H3,(H,2,3,4)/q+1;/p-1/b9-7+;

InChI key

ZXQHSPWBYMLHLB-BXTVWIJMSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fumika Mi-ichi et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(22), E2884-E2890 (2015-05-20)
Hydrogenosomes and mitosomes are mitochondrion-related organelles (MROs) that have highly reduced and divergent functions in anaerobic/microaerophilic eukaryotes. Entamoeba histolytica, a microaerophilic, parasitic amoebozoan species, which causes intestinal and extraintestinal amoebiasis in humans, possesses mitosomes, the existence and biological functions of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service