Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

805076

Sigma-Aldrich

2-(Trimethylsilyl)cyclopent-2-en-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14OSi
CAS Number:
Molecular Weight:
154.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

refractive index

n20/D 1.472

transition temp

flash point 165 °F

density

0.936 g/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

O=C1CCC=C1[Si](C)(C)C

InChI

1S/C8H14OSi/c1-10(2,3)8-6-4-5-7(8)9/h6H,4-5H2,1-3H3

InChI key

JREFDYKOYRJDQC-UHFFFAOYSA-N

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

165.0 °F

Flash Point(C)

73.88 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service