Skip to Content
MilliporeSigma
All Photos(2)

Documents

804940

Sigma-Aldrich

5-Bromo-2-benzofuranboronic acid MIDA ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11BBrNO5
Molecular Weight:
351.95
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

SMILES string

BrC1=CC=C2C(C=C([B-]3([N+](C4)(C)CC(O3)=O)OC4=O)O2)=C1

InChI

1S/C13H11BBrNO5/c1-16-6-12(17)20-14(16,21-13(18)7-16)11-5-8-4-9(15)2-3-10(8)19-11/h2-5H,6-7H2,1H3

InChI key

ZNACQWWAWVGCRP-UHFFFAOYSA-N

General description

5-Bromo-2-benzofuranboronic acid MIDA ester is a MIDA (N-methyliminodiacetic acid) protected boronate ester. Some of its unique features include air-stability, compatibility with common harsh synthetic reagents, solubility in various organic solvents and compatibility with silica-gel. These attributes makes it highly useful in Suzuki-Miyaura cross-coupling reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service