Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

804835

Sigma-Aldrich

P(o-tol)3 Pd G3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H34NO3PPdS
CAS Number:
Molecular Weight:
674.10
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

form

powder

feature

generation 3

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

functional group

phosphine

SMILES string

NC1=C(C2=CC=CC=C2[Pd]OS(C)(=O)=O)C=CC=C1.CC3=CC=CC=C3P(C4=CC=CC=C4C)C5=CC=CC=C5C

InChI

1S/C21H21P.C12H10N.CH4O3S.Pd/c1-16-10-4-7-13-19(16)22(20-14-8-5-11-17(20)2)21-15-9-6-12-18(21)3;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h4-15H,1-3H3;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

LUGPLNLXIADYMW-UHFFFAOYSA-M

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

G3 and G4 Buchwald palladium precatalysts are the newest air, moisture, and thermally stable crossing-coupling complexes used in bond formation for their versatility and high reactivity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service