Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

803111

Sigma-Aldrich

25-Hydroxyvitamin D3-26,26,26,27,27,27-d6 sulfate sodium salt solution

100 μg/mL in ethanol, 98 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27D6H37O5SNa
CAS Number:
Molecular Weight:
508.72
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

97% (CP)

form

solution

concentration

100 μg/mL in ethanol

technique(s)

mass spectrometry (MS): suitable

color

colorless

application(s)

detection

mass shift

M+6

storage temp.

−20°C

SMILES string

C[C@H](CCCC(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)C1CCC([C@]1(C)CCC/2)C2=C\C=C(C[C@@H](OS(=O)([O-])=O)CC3)/C3=C.[Na+]

InChI

1S/C27H44O5S.Na/c1-19-10-13-23(32-33(29,30)31)18-22(19)12-11-21-9-7-17-27(5)24(14-15-25(21)27)20(2)8-6-16-26(3,4)28;/h11-12,20,23-25,28H,1,6-10,13-18H2,2-5H3,(H,29,30,31);/q;+1/p-1/b21-11+,22-12-;/t20-,23+,24?,25?,27-;/m1./s1/i3D3,4D3;

InChI key

UVFUTTRHHIQISA-DZAJWRDGSA-M

Related Categories

General description

25-Hydroxyvitamin D3 (calcifediol) is a prohormone produced via hydroxylation of vitamin D3 (cholecalciferol) in the liver. It is a precursor for the synthesis of calcitriol {1,25-dihydroxyvitamin D3 or [1,25(OH)2D3]}. It is used as a biomarker to determine the status of vitamin D in the body.[1][2]
25-Hydroxyvitamin D3-26,26,26,27,27,27-d6 sulfate sodium salt is a deuterated vitamin D3 wherein methyl protons of C-26, C-27 carbons are replaced by deuterium.

Application

25-Hydroxyvitamin D3-26,26,26,27,27,27-d6 sulfate sodium salt can be used as a stable isotope internal standard for data interpretation in specific biological samples by mass spectrometry.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of circulating 25-hydroxyvitamin D by liquid chromatography-tandem mass spectrometry
Vogeser M
The Journal of Steroid Biochemistry and Molecular Biology, 121(3-5), 565-573 (2010)
25-Hydroxyvitamin D3, the prohormone of 1, 25-dihydroxyvitamin D3, inhibits the proliferation of primary prostatic epithelial cells
Barreto, AM et al.
Cancer Epidemiology, Biomarkers & Prevention, 9(3), 265-270 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service