Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

803065

Sigma-Aldrich

Progesterone-2,3,4-13C3 solution

100 μg/mL in acetonitrile, 99 atom % 13C, 98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$1,110.00

$1,110.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 ML
$1,110.00

About This Item

Empirical Formula (Hill Notation):
13C3C18H30O2
Molecular Weight:
317.44
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12

$1,110.00


Please contact Customer Service for Availability

Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

98% (CP)

form

solution

concentration

100 μg/mL in acetonitrile

technique(s)

mass spectrometry (MS): suitable

mass shift

M+3

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@@]12C(CC[C@@H]2C(C)=O)C3CCC4=[13CH][13C]([13CH2]C[C@]4(C)C3CC1)=O

InChI

1S/C21H30O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12,16-19H,4-11H2,1-3H3/t16?,17-,18?,19?,20+,21-/m1/s1/i8+1,12+1,15+1

InChI key

RJKFOVLPORLFTN-LLYMGNADSA-N

Related Categories

Application

This product may be used as an analytical standard.

Packaging

1 mL in ampule
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service