Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

802646

Sigma-Aldrich

Aquivion® PW87S

coarse powder, PFSA eq. wt. 870 g/mole SO3H, contains CF3 polymer chain ends as stabilizer

Synonym(s):

Aquivion® SO3H, Tetrafluoroethylene-perfluoro(3-oxa-4-pentenesulfonic acid) copolymer, Ethanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CF2CF(OCF2CF2SO3H)]m[CF2CF2]n
CAS Number:
UNSPSC Code:
26111700
NACRES:
NA.23

form

coarse powder

contains

CF3 polymer chain ends as stabilizer

Looking for similar products? Visit Product Comparison Guide

General description

Aquivion® PFSA PW series grades are coarse powders based on the short-side-chain (SSC) copolymer of Tetrafluoroethylene and the Sulfonyl Fluoride Vinyl Ether (SFVE) CF2=CF2-O-(CF2)2-SO2F industrially produced by Solvay Specialty Polymers. The resulting perfluoropolymer′s functional groups are in their sulfonic acid form, SO3H.

Application

  • Aquivion® PW87S acid powder exhibits ion cation-exchange capacity and can be used as such under conditions where conventional exchange resins have shown insufficient chemical resistance.
  • Given the resin′s superacid functionality, it can also be used either directly or as an ingredient of composite structures in the field of heterogeneous catalysis. This encompasses a wide range of organic synthesis and respective reaction mechanisms, as already described in public literature.

Legal Information

Product of Solvay Specialty Polymers USA LLC.
Aquivion is a registered trademark of Syensqo Group

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Advances in electrochemical water conversion and understanding PEMFC degradation drive progress in hydrogen technologies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service