Skip to Content
MilliporeSigma
All Photos(1)

Documents

80050

Sigma-Aldrich

Bis(methylglycol) phthalate

technical, ≥90% (GC)

Synonym(s):

Bis(2-methoxyethyl) phthalate, Dimethylglycol phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18O6
CAS Number:
Molecular Weight:
282.29
Beilstein:
2056929
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥90% (GC)

form

liquid

refractive index

n20/D 1.503

bp

230 °C/10 mmHg (lit.)

density

1.173 g/mL at 20 °C (lit.)

SMILES string

COCCOC(=O)c1ccccc1C(=O)OCCOC

InChI

1S/C14H18O6/c1-17-7-9-19-13(15)11-5-3-4-6-12(11)14(16)20-10-8-18-2/h3-6H,7-10H2,1-2H3

InChI key

HSUIVCLOAAJSRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M R Parkhie et al.
Environmental health perspectives, 45, 89-97 (1982-11-01)
A single intraperitoneal injection (0.6 ml/kg) of dimethoxyethyl phthalate (DMEP) was given to groups of Wistar strain rats on day 10, 11, 12, 13 or 14 of gestation. Control rats received 0.6 ml/kg of physiological saline intraperitoneally. In phthalate-treated rats
J Campbell et al.
Journal of applied toxicology : JAT, 4(1), 35-41 (1984-02-01)
The rat foetus, in contrast with the maternal liver and placenta, has little or no ability to hydrolyse di-(2-methoxyethyl)-phthalate (DMEP) to mono-2-methoxyethyl)-phthalate (MMEP). At short times after the administration of DMEP to the dam on the 14th day of gestation
R Gollamudi et al.
Journal of applied toxicology : JAT, 5(6), 368-371 (1985-12-01)
Di(2-ethylhexyl)phthalate (DEHP) inhibited UDP-glucuronyltransferase activity of rat liver in vitro and in vivo. Diethyl phthalate and dimethoxyethyl phthalate also inhibited this enzyme in vitro. On the other hand, DEHP did not inhibit the activity of the cytosolic enzyme N-acetyltransferase; it
Wataru Naito et al.
Environmental monitoring and assessment, 115(1-3), 451-471 (2006-05-02)
Screening-level ecological risk assessments of di(2-ethylhexyl)phthalate (DEHP) for aquatic organisms in Japan were conducted using estimated statistical values based on surface water and sediment monitoring data and effect threshold values based on a large aquatic toxicity database. An alternative method
S L Cassidy et al.
Archives of toxicology, 53(1), 71-78 (1983-05-01)
Single dosages of DMEP (1,000-2,000 mg/kg), GMCH (50 mg/kg), ECH (25 and 50 mg/kg), FA (100 and 200 mg/kg), and MMS (100-400 mg/kg) were administered orally to 10 week old male Wistar rats. The rats were necropsied on the 11th

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service