Skip to Content
MilliporeSigma
All Photos(2)

Documents

799297

Sigma-Aldrich

1,1,2,2,9,9,10,10-Octafluoro[2.2]paracyclophane

Synonym(s):

α-Perfluorodi-p-xylene, 1,1,2,2,9,9,10,10-Octafluoro-p-cyclophane, 2,2,3,3,8,8,9,9-octafluorotricyclo[8.2.2.24,7]hexadeca-4,6,10,12,13,15-hexaene, Parylene AF 4 dimer

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H8F8
CAS Number:
Molecular Weight:
352.22
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.23

Assay

99% (GC)

Quality Level

form

powder

mp

240-250 °C

SMILES string

FC1(F)C2=CC=C(C(F)(F)C(F)(F)C3=CC=C(C1(F)F)C=C3)C=C2

InChI

1S/C16H8F8/c17-13(18)9-1-2-10(4-3-9)14(19,20)16(23,24)12-7-5-11(6-8-12)15(13,21)22/h1-8H

InChI key

KCKIWSAAWFKXMA-UHFFFAOYSA-N

General description

Characteristics of Parylen AF4 coating:
Specialties:
  • Unmatched UV stability
  • Excellent thermal stability at high service temperature (350)
  • Unmatched oxidation stability
  • Completely conformal coating
  • Superior crevice penetration, pin hole free
  • Anti-saline damage
  • Anti-stick ion

Barrier properties:
  • Chemical insolubility and solvent resistant
  • Low water absorption
  • Hydrophobic properties
  • Low gas permeability , no out-gassing

Physical & mechanical properties:
  • Transparent
  • Light-weight and ultra-thin
  • Low CTE
  • Low coefficient of friction
  • Good dry lubricity
  • Abrasion resistance
  • Stabilization of components and structures
  • Particulate immobilization

Electrical properties:
  • Electrical insulation
  • High dielectric strength
  • High volume resistivity ,ohm-cm
  • Low dissipation factor
  • Low dielectric constant
  • Mitigation of tin whisker growth

Biocompatibility and bio-stability
  • USP class VI
  • Microorganism resistance

Application

Parylen AF4 is extremely useful in high temperature applications and those in which long-term UV stability is required.
  • Aerospace
  • Military
  • Electronic
  • Medical
  • Industrial
  • Semiconductor (3D IC TSV electrical insulation , AMOLED De-bonding layer , Barrier layer )
  • Bio-MEMS electrode protection film
  • EWD (Electro Wetting Device) Liquid lens

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service