Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

799017

Sigma-Aldrich

Carbon nanochips

Synonym(s):

CNC, CNCF, Heat treated platelet graphite nanofibers, carbon nanochip fibers

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

flakes

size

, aspect Ratio: 25
, average length: 2.5μ
, average width : 100 nm

surface area

40 m2/g

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

General description

Carbon nanochips consist of wrapped graphite sheets oriented perpendicular to the fiber axis. Carbon nanochips are formed when platelet graphene or graphite nanofibers are heated in argon to about 3000°C, resulting in a sealing action of some of the edge sites and also removes all traces of inclusions such as the catalyst that is used to grow the precursor graphite nanofibers.

Application

  • Negative electrode for lithium ion secondary batteries.
  • Catalyst for ethylene oxidative dehydrogenation.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel carbon nanofibers of high graphitization as anodic materials for lithium ion secondary batteries
Yoon SH, .
Carbon, 42, 21-32 (2003)
Ethylene oxidation over Ag supported on novel carbon nano-structured supports
Xu X,
Reaction Kinetics and Catalysis Letters, 87, 305-312 (2006)
Carbon nanochips and nanotubes decorated PtAuPd-based nanocomposites for glucose sensing: Role of support material and efficient Pt utilisation
Singh B,
Sensors and Actuators B, Chemical, 205, 401-410 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service