Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

798819

Sigma-Aldrich

(S)-N-Butyl-1-[(S)-2-((E)-2-hydroxybenzylideneamino)-3-methylbutanoyl]pyrrolidine-2-carboxamide

Synonym(s):

Yoon Anti-[2+2]-Photocycloaddition Ligand

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H31N3O3
CAS Number:
Molecular Weight:
373.49
UNSPSC Code:
12352111
PubChem Substance ID:

form

powder

reaction suitability

reaction type: Asymmetric synthesis
reagent type: catalyst
reaction type: Photocatalysis

functional group

amide
amine
imine

storage temp.

2-8°C

SMILES string

OC1=C(/C=N/[C@@H](C(C)C)C(N2[C@H](C(NCCCC)=O)CCC2)=O)C=CC=C1

InChI

1S/C21H31N3O3/c1-4-5-12-22-20(26)17-10-8-13-24(17)21(27)19(15(2)3)23-14-16-9-6-7-11-18(16)25/h6-7,9,11,14-15,17,19,25H,4-5,8,10,12-13H2,1-3H3,(H,22,26)/b23-14+/t17-,19-/m0/s1

InChI key

DULOQBOELOOGIJ-DYOPXDPWSA-N

Application

Chiral ligand for the asymmetric [2+2] cycloaddition of enones by photoredox catalysis.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Yoon lab exploits visible light photochemistry for unique chemical transformations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service