Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

798460

Sigma-Aldrich

2,7-Dimethyl-10-phenylacridin-9(10H)-one

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H17NO
CAS Number:
Molecular Weight:
299.37
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

functional group

ketone

SMILES string

CC1=CC=C(N(C2=CC=CC=C2)C(C=CC(C)=C3)=C3C4=O)C4=C1

InChI

1S/C21H17NO/c1-14-8-10-19-17(12-14)21(23)18-13-15(2)9-11-20(18)22(19)16-6-4-3-5-7-16/h3-13H,1-2H3

InChI key

ZFGJBTQJOSUQPV-UHFFFAOYSA-N

Application

Building block was used as a key intermediate in the synthesis of acridinium catalyst developed by Nicewicz and coworkers, Aldrich 793876.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Nicewicz lab focuses on reactive cation radical species for selective reactions, advancing synthesis methodologies for various functional groups.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service