Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

797561

Sigma-Aldrich

7-Hydroxy-5-thia-2-azaspiro[3.4]octane-2-carboxylic acid-5,5-dioxide 1,1-dimethylethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO5S
CAS Number:
Molecular Weight:
277.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

functional group

hydroxyl
sulfone

storage temp.

2-8°C

SMILES string

OC(CS1(=O)=O)CC21CN(C(OC(C)(C)C)=O)C2

InChI

1S/C11H19NO5S/c1-10(2,3)17-9(14)12-6-11(7-12)4-8(13)5-18(11,15)16/h8,13H,4-7H2,1-3H3

InChI key

ZKCKSGZYCIBCKJ-UHFFFAOYSA-N

Application

Spirocyclic building block was first synthesized by Carreira and coworkers, which provides a new area of chemical space with straightforward functional handles for further diversification.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Spirocyclic modules containing four-membered rings are currently of growing interest to discovery chemists.

Related Content

Carreira Research & SpiroChem innovate spirocyclic building blocks, enhancing drug discovery with diverse functionalization sites and compact modules.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service