Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

797383

Sigma-Aldrich

Sodium 3,3′-{[(1,2-diphenylethene-1,2-diyl)bis(4,1-phenylene)]bis(oxy)}bis(propane-1-sulfonate)

Synonym(s):

TPE-Sulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H30Na2O8S2
CAS Number:
Molecular Weight:
652.69
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

SMILES string

[O-]S(CCCOC(C=C1)=CC=C1/C(C2=CC=CC=C2)=C(C3=CC=C(OCCCS([O-])(=O)=O)C=C3)\C4=CC=CC=C4)(=O)=O.[Na+].[Na+]

InChI

1S/C32H32O8S2.2Na/c33-41(34,35)23-7-21-39-29-17-13-27(14-18-29)31(25-9-3-1-4-10-25)32(26-11-5-2-6-12-26)28-15-19-30(20-16-28)40-22-8-24-42(36,37)38;;/h1-6,9-20H,7-8,21-24H2,(H,33,34,35)(H,36,37,38);;/q;2*+1/p-2/b32-31+;;

InChI key

ZIQVFZHFAUZKPD-GLDAUVFXSA-L

Looking for similar products? Visit Product Comparison Guide

Application

TPE-Sulfonate is an aggregation-induced emission (AIE) dye for bio-imaging and water-based fluorescent probes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuning Hong et al.
Analytical chemistry, 82(16), 7035-7043 (2010-08-14)
Human serum albumin (HSA) is a major protein component of blood plasma, and its assay is of obvious value to biological research. We, herein, present a readily accessible fluorescent bioprobe for HSA detection and quantitation. A nonemissive tetraphenylethene derivative named

Articles

AIE Luminogens - Family of new materials with multifaceted functionalities. Find some representative applications of the AIEgens

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service