Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

797332

Sigma-Aldrich

1,2-Bis[4-(bromomethyl)phenyl]-1,2-diphenylethene

Synonym(s):

1,1′-(1,2-Diphenyl-1,2-ethenediyl)bis[4-(bromomethyl)benzene], TPE-MB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H22Br2
CAS Number:
Molecular Weight:
518.28
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

SMILES string

BrCC(C=C1)=CC=C1/C(C2=CC=CC=C2)=C(C3=CC=CC=C3)/C4=CC=C(CBr)C=C4

InChI

1S/C28H22Br2/c29-19-21-11-15-25(16-12-21)27(23-7-3-1-4-8-23)28(24-9-5-2-6-10-24)26-17-13-22(20-30)14-18-26/h1-18H,19-20H2/b28-27+

InChI key

GHZTXESUSOBGAM-BYYHNAKLSA-N

Application

TPE-MB is an intermediate of aggregation-induced emission (AIE) material used in the synthesis of blue mitochondrial dyes for bio-imaging. TPE-MB may be used in the preparation of aggregation induced emission (AIE) probes for the fluorescence detection of mercury(II) and glutathione. [1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Label-free fluorescence detection of mercury (II) and glutathione based on Hg2+-DNA complexes stimulating aggregation-induced emission of a tetraphenylethene derivative.
Xu JP,et al.
Analyst, 3002-3007 null
Chris Wai Tung Leung et al.
Journal of the American Chemical Society, 135(1), 62-65 (2012-12-19)
Tracking the dynamics of mitochondrial morphology has attracted much research interest because of its involvement in early stage apoptosis and degenerative conditions. To follow this process, highly specific and photostable fluorescent probes are in demand. Commercially available mitochondria trackers, however
Label-free fluorescence detection of mercury (II) and glutathione based on Hg2+-DNA complexes stimulating aggregation-induced emission of a tetraphenylethene derivative.
Xu JP,et al.
Analyst null

Articles

AIE Luminogens - Family of new materials with multifaceted functionalities. Find some representative applications of the AIEgens

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service