Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

797286

Sigma-Aldrich

2,5-Bis[4-(azidomethyl)phenyl]-1,1-dimethyl-3,4-diphenyl-1H-silole

96%

Synonym(s):

1,1-Dimethyl-2,5-bis[4-(azidomethyl)phenyl]-3,4-diphenylsilole, 2,5-Bis[4-(azidomethyl)phenyl]-1,1-dimethyl-3,4-diphenylsilacyclopenta-2,4-diene, Silole-N3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H28N6Si
CAS Number:
Molecular Weight:
524.69
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

form

solid

SMILES string

C[Si]1(C)C(C2=CC=C(CN=[N+]=[N-])C=C2)=C(C3=CC=CC=C3)C(C4=CC=CC=C4)=C1C5=CC=C(CN=[N+]=[N-])C=C5

InChI

1S/C32H28N6Si/c1-39(2)31(27-17-13-23(14-18-27)21-35-37-33)29(25-9-5-3-6-10-25)30(26-11-7-4-8-12-26)32(39)28-19-15-24(16-20-28)22-36-38-34/h3-20H,21-22H2,1-2H3

InChI key

ZMHVJCSUTIHLNO-UHFFFAOYSA-N

General description

Silole is an represents a species of efficient emitter. A variety of applications can be developed through functionalization of silole. Taking advantage of the azide-alkyne click reaction, silole can be easily tailored to construct different materials.

Application

Silole-N3 is a "Clickable" aggregation-induced emission (AIE) material for use in circularly polarised luminescence (CPL).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

What makes efficient circularly polarised luminescence in the condensed phase: aggregation-induced circular dichroism and light emission
Jianzhao Liu, Huimin Su, Luming Meng, Yihua Zhao, Chunmei Deng, Jason C. Y. Ng, et al
Chemical Science, 3, 2737-2747 (2013)

Articles

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service