Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

797022

Sigma-Aldrich

tert-Butyl 3,3-difluoro-4-oxopiperidine-1-carboxylate hydrate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17F2NO4
CAS Number:
Molecular Weight:
253.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

mp

102-107 °C

functional group

fluoro
ketone

SMILES string

O.CC(C)(C)OC(=O)N1CCC(=O)C(F)(F)C1

InChI

1S/C10H15F2NO3.H2O/c1-9(2,3)16-8(15)13-5-4-7(14)10(11,12)6-13;/h4-6H2,1-3H3;1H2

InChI key

IEESRSKOIOREKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Spirocyclic modules containing four-membered rings are currently of growing interest to discovery chemists.

Related Content

Carreira Research & SpiroChem innovate spirocyclic building blocks, enhancing drug discovery with diverse functionalization sites and compact modules.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service