Skip to Content
MilliporeSigma
All Photos(3)

Documents

796247

Sigma-Aldrich

Silver nanospheres

100 nm avg. part. size, 0.02 mg/mL in water, lipoic acid functionalized

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ag
Molecular Weight:
107.87
UNSPSC Code:
12352300
NACRES:
NA.23

form

nanospheres

concentration

0.02 mg/mL in water

avg. part. size

100 nm

density

1.0005 g/mL at 25 °C

λmax

485 nm

functional group

lipoic acid

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Store products away from light at 4-6°C. Short periods at room temperature are acceptable however, lower temperature storage prolongs the shelf life of the product. DO NOT FREEZE.

Application

Silver nanomaterials have unique physical, chemical and optical properties that are currently being leveraged for a wide variety of biological applications. Additionally, silver nanoparticles have an optical color that is a function of the silver nanoparticle’s size and shape. The strong coupling of silver nanoparticles to specific wavelengths of incident light can be utilized to develop ultrabright reporter molecules, highly efficient thermal absorbers, and nanoscale "antennas" that can amplify the strength of the local electromagnetic field.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Research on noble bimetallic nanoparticles for glucose biosensor detection.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service