Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

796107

Sigma-Aldrich

Cerium iron oxide hydroxide

aqueous nanoparticle dispersion, <5 nm (DLS), 20% solids by weight, pH ~4.75

Synonym(s):

Cerium iron mixed metal oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CeFeHO4
Molecular Weight:
260.97
UNSPSC Code:
12352302
NACRES:
NA.23

form

dispersion

reaction suitability

reagent type: catalyst
core: cerium

particle size

<5 nm (DLS)

pH

~4.75

density

1.23 g/mL at 25 °C

storage temp.

2-8°C

Application

This material is highly monodisperse (<0.25 polydispersity index) in size and shape, and provides significantly improved surface reactivity. It is stable at high suspension densities, when utilized at the recommended pH range (4.5 – 5.0). Applications include emissions catalysis, regeneration of particle traps in diesel engines and oxidation catalysis.

Legal Information

Product of Cerion, Inc

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service