Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

795151

Sigma-Aldrich

(S)-t-Bu-Quinox

97%

Synonym(s):

2-[(4S)-4-(1,1-dimethylethyl)-4,5-dihydro-2-oxazolyl]quinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N2O
CAS Number:
Molecular Weight:
254.33
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

97%

form

solid

reaction suitability

reaction type: Cross Couplings

mp

93-98 °C

SMILES string

CC(C)(C)[C@@H](CO1)N=C1C2=NC3=CC=CC=C3C=C2

InChI

1S/C16H18N2O/c1-16(2,3)14-10-19-15(18-14)13-9-8-11-6-4-5-7-12(11)17-13/h4-9,14H,10H2,1-3H3/t14-/m1/s1

InChI key

ZECIAWBAXVMXKI-CQSZACIVSA-N

Application

Ligand has been shown to influence high enantioselectivity in the asymmetric conjugate addition of boronic acids to enones under palladium catalysis.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service